Seward BA6040 Stomacher® 80 Biomaster Standard Bags, Square Bottom; 2000/CS
Representative image only.
Seward BA6040 Stomacher® 80 Biomaster Standard Bags, Square Bottom; 2000/CS
Seward | Mfr: BA6040| Item: 3685091
Eliminates the risk of crosss contamination
Call For Price
Call For Price
Buy It Together
Current Item
Seward BA6040 Stomacher® 80 Biomaster Standard Bags, Square Bottom; 2000/CS
Seward BA6040 Stomacher® 80 Biomaster Standard Bags, Square Bottom; 2000/CS
3685091
Call For Price
+ +
Seward BA6090 Blending Bag Rack, Holds 10 Bags
3686002
Call For Price
=
Total Price 0 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
  • Manufactured from high-grade, food quality polythene
  • Minimum 65 micron film thickness provides high-impact strength
  • All Seward Stomacher Bags are manufactured from virgin LDPE polymer. The only seal is at the base.
  • Compatible with all 80 lab blender homogenizers.
  • All Seward Stomacher Bags are gamma irradiated at 25 to 50kGy to guarantee sterility.
  • 1 year warranty
More About this Item
Choose from a variety of bags to use with your lab blender. Blending bags are made from high quality food grade nontoxic virgin LDPE granule, extruded to 65µm film thickness. All blender bags are irradiated sterile. A certificate or irridiation is available upon request.
The Stomacher® 80 bag has been designed to fulfill all applications from blending, straining and storage including blender bags designed specifically for life science applications.
1 Year Warranty
Specifications
  • Bag TypePaddle Blender Bags
  • Bag Capacity (mL)80
  • Bag Capacity (L)0.08
  • DescriptionStomacher bags; use with Stomacher 80, square bottom, 2000/cs