Setra SRAN Remote Annunciator for Visual and Audible Status Indication
Representative image only.
Setra SRAN Remote Annunciator for Visual and Audible Status Indication
Setra | Mfr: SRAN| Item: 9807490
Optimizer performance of your pressure transducer
₹88,500.00INR / Each
₹88,500.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
These accessories are compatible with the Setra 98075-XX series AccuSense™ High-Accuracy Pressure Transducers.
The SecureCal™ calibrator 98075-95 is used to calibrate transducers in the field—enables zero-tare and snap-span adjustment, factory calibration restore function, and two-line LCD/LED feedback.
1 Year Warranty
Specifications
  • DescriptionRemote Annunciator for Visual and Audible Status Indication