Sensorex FC75P Conductivity Probe, K=10 with ATC-P1K, 3/4"NPT(M)
Representative image only.
Sensorex FC75P Conductivity Probe, K=10 with ATC-P1K, 3/4"NPT(M)
Sensorex | Mfr: CS200TC-K=10 + FC75P| Item: 1906618
Ideal for laboratory or light industrial use
₹61,000.00INR / Each
₹61,000.00INR / Each
Product Description
Key Features
  • Graphite measuring surfaces
  • PT 1000 RTD temperature compensation
  • 3/4" M NPT mounting colar
  • 1 year warranty
More About this Item
Economical laboratory or light industrial conductivity sensors feature either graphite or platinum ring technology. The durable epoxy body construction provides a rugged, dependable sensor. Suitable for most conductivity meters.
1 Year Warranty
Specifications
  • Cell ConstantK = 10
  • Cell TypeIn-line, submersible
  • Electrode MaterialGraphite
  • Body MaterialEpoxy
  • Fitting¾" NPT(M)
  • Mounting TypeThreaded
  • Cable Length (m)3
  • Cable Length (ft)10
  • DescriptionConductivity Probe, K=10 with ATC-P1K, 3/4"NPT(M)