Sensorex 971651 In-line Dissolved Oxygen Probe for DO Transmitters and Controllers
Representative image only.
Sensorex 971651 In-line Dissolved Oxygen Probe for DO Transmitters and Controllers
Sensorex | Mfr: 971651| Item: 3520156
Designed to measure partial pressure of oxygen in the water
₹80,500.00INR / Each
₹80,500.00INR / Each
Buy It Together
Current Item
Sensorex 971651 In-line Dissolved Oxygen Probe for DO Transmitters and Controllers
Sensorex 971651 In-line Dissolved Oxygen Probe for DO Transmitters and Controllers
3520156
₹80,500.00INR / Each
+ =
Total Price 99100.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Ideal for use with Oakton oxygen DO instruments
  • 1 year warranty
More About this Item
Probe assembly consists of a 12 mm body, galvanic dissolved oxygen electrode with a 3/4" NPT(M) compression fitting. Probe has 10 feet of cable with tinned end leads and 100 Ohm Pt RTD temperature compensation.
Install into a 3/4" tee fitting (not included) so that fluid is in constant contact with probe membrane.
1 Year Warranty
Specifications
  • Meter TypeAccessory; Probes and Sensors
  • Min Temperature (° C)0
  • Max Temperature (° C)50
  • Min Temperature (° F)32
  • Max Temperature (° F)122
  • Temperature Accuracy+2% when measuring temp equal cal temp.
  • Output26 mV ±8 millivolts
  • DescriptionIn-line Dissolved Oxygen Probe for DO Transmitters and Controllers