Secador 5.0 Desiccator Shelf

Representative image only.

Secador 5.0 Desiccator Shelf

Secador | Mfr: 420571001| Item: 0500628

Easily add additional self space to your desiccator cabinet

₹28,700.00INR / Each

₹28,700.00INR / Each
Product Description

Key Features

  • Fits Secador 5.0 desiccator cabinets
  • Three month warranty

More About this Item

This clear polystyrene shelf offers additional space to store products in your existing Secador® 5.0 desiccator cabinet.

3 Month Warranty
Specifications
  • Interior Width (in)11
  • Interior Depth (in)13.385
  • Interior Width (cm)28
  • Interior Depth (cm)34
  • MaterialPolystyrene
  • DescriptionDesiccator Shelf
  • Warranty3 months
Reviews
Q&A