Scienceware Magnetic Stir Bar Kit with Spinning Ring; 10 Bars
Representative image only.
Scienceware Magnetic Stir Bar Kit with Spinning Ring; 10 Bars
Scienceware | Mfr: F37158-0010| Item: 0477597
Simplify your stirring with these kits
₹18,200.00INR / Each
₹18,200.00INR / Each
Product Description
Includes
Stir bar retainer and two each 1/2" x 1/8”, 1/2" x 5/16”, 1" x 5/16”, 1 1/2" x 5/16”, and 2" x 5/16” sizes.
Key Features
  • Chemically inert
  • Low friction
  • Leakproof PTFE coating
  • Stir bar retainer holds the stir bar in the vessel while pouring
  • 3 month warranty
More About this Item
Kits each offer a wide variety of stir bar sizes to accomodate different glassware in the lab.
Features 10 of the most common stir bar sizes in one convenient kit. Each Alnico magnet is coated in inert PTFE. Comes with a variety of sizes, and colors, to thoroughly mix small and large volumes
3 Month Warranty
Specifications
  • DescriptionSpinpak Magnetic Stirring Bar Assortment; 10 Stir Bars