Scienceware H37801-0000 Acid/Caustic Solution Sampler, 16 oz capacity, 36" handle
Representative image only.
Scienceware H37801-0000 Acid/Caustic Solution Sampler, 16 oz capacity, 36" handle
Scienceware | Mfr: H37801-0000| Item: 3368770
Safely take samples of caustic solutions from drums, tanks, and barges
₹80,000.00INR / Each
₹80,000.00INR / Each
Product Description
Key Features
  • Ideal for sampling from large containers and reservoirs
More About this Item
These polyethylene samplers are designed for taking samples of caustic solutions like sodium hydroxide, potassium hydroxide, hydrocloric acid, acetic acid, formaldehyde, and sulfic acid.
Specifications
  • DescriptionAcid/Caustic Solution Sampler, 16 oz capacity, 36" handle