Scienceware H36719-0000 Stainless Steel Vibrating Spatula with PTFE Coating
Representative image only.
Scienceware H36719-0000 Stainless Steel Vibrating Spatula with PTFE Coating
Scienceware | Mfr: H36719-0000| Item: 0626455
Maneuvers easily into small-mouth containers and tight corners
₹34,000.00INR / Pkg of 1
₹34,000.00INR / Pkg of 1
Product Description
Key Features
  • Simply spin thumbwheel to control the amount added
  • Ideal for use with analytical balance
More About this Item
Move the toothed thumbwheel to “vibrate” the blade—lets you add as little or as much of your precious material as needed. Narrow V-shaped blade maneuvers easily into small-mouth containers and tight corners. Stainless steel blade is also available with a brown PTFE coating; FDA compliant for direct food contact applications.
Specifications
  • Overall Length (in)8 1/4
  • Overall Length (cm)20.96
  • MaterialStainless Steel with PTFE coating
  • SterileNo
  • DescriptionStainless steel vibrating spatula with PTFE coating