Scienceware H18901-0000 Microcentrifuge Tube Ice Rack (holds 105 X 400 UL Tubes)
Representative image only.
Scienceware H18901-0000 Microcentrifuge Tube Ice Rack (holds 105 X 400 UL Tubes)
Scienceware | Mfr: H18901-0000| Item: 0627409
For temperature stabilized benchtop ice storage
₹39,300.00INR / Each
₹39,300.00INR / Each
Product Description
More About this Item
These two piece racks are molded of high-impact polystyrene with a lower section tray that holds water or ice flakes.
Holds 105 400 µl microcentrifuge tubes, or other tubes with a diameter up to 8 mm.
Specifications
  • Tubes Held105
  • MaterialPolypropylene(PP)
  • DescriptionMicrocentrifuge Tube Ice Rack (holds 105 X 400 UL Tubes)