Scienceware F371500000 Spinbar,cell Stirring Bar,ptfe 9 X 8 Mm
Representative image only.
Scienceware F371500000 Spinbar,cell Stirring Bar,ptfe 9 X 8 Mm
Scienceware | Mfr: F371500000| Item: 0477519
₹5,500.00INR / Each
₹5,500.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Specifically for Spectrophotometer cells or cuvettes, this one-piece molded stir bar fits into standard 10mm light path cuvette. Provides rapid vertical and horizontal mixing with minimum vortexing when placed on a stirrer. Photometric measurements can proceed uninterrupted while reagents are added.
1 Year Warranty
Specifications
  • Diameter (in)0.315
  • Diameter (cm)0.8
  • DescriptionSpinbar,cell Stirring Bar,ptfe 9 X 8 Mm