Scienceware F371460250 Spinstar Stirring Bar, PTFE, 2-7/16"
Representative image only.
Scienceware F371460250 Spinstar Stirring Bar, PTFE, 2-7/16"
Scienceware | Mfr: F371460250| Item: 0477508
₹16,400.00INR / Each
₹16,400.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Star-shape creates deep vortex at slow speeds. Fits exactly inside diameter of 250 mL beaker.
1 Year Warranty
Specifications
  • DescriptionSpinstar Stirring Bar, PTFE, 2-7/16"