Scienceware F371400010 Spinring Stirring Bar, PTFE, 1-1/4"od
Representative image only.
Scienceware F371400010 Spinring Stirring Bar, PTFE, 1-1/4"od
Scienceware | Mfr: F371400010| Item: 0477542
₹12,600.00INR / Each
₹12,600.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Removable stabilizing ring keeps stir bar centered in vessel. Octagon shape creates intense vortex without splashing or chattering and resists spinout.
1 Year Warranty
Specifications
  • Diameter (in)0.25
  • Diameter (cm)0.635
  • DescriptionSpinring Stirring Bar, PTFE, 1-1/4"od