Scienceware F371250014 Spinfin Stirring Bar, PTFE, 5/16" X 3/8"
Representative image only.
Scienceware F371250014 Spinfin Stirring Bar, PTFE, 5/16" X 3/8"
Scienceware | Mfr: F371250014| Item: 0477529
₹2,600.00INR / Each
₹2,600.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Vigorously shakes liquids in round-bottom test tubes and flasks, where other stir bars cannot be used. Position is maintained in a vessel and will not vibrate.
1 Year Warranty
Specifications
  • Diameter (in)0.375
  • Diameter (cm)0.9525
  • DescriptionSpinfin Stirring Bar, PTFE, 5/16" X 3/8"