Scienceware F36721-0025 Polystyrene Sampling Spoons, 0.25 mL Capacity; 25/Pk

Representative image only.

Scienceware F36721-0025 Polystyrene Sampling Spoons, 0.25 mL Capacity; 25/Pk

Scienceware | Mfr: F36721-0025| Item: 0626354

Reduce cross-contamination

₹10,800.00INR / Each

₹10,800.00INR / Each
Product Description

Key Features

  • Ideal for single use
  • Made of high-impact polystyrene

More About this Item

High-impact polystyrene sampling spoons are precise enough to collect specific volume amounts and economical enough to use a separate spoon for each substance — reducing your chances of cross-contamination.

Specifications
  • Capacity (oz)0.0085
  • Capacity (mL)0.25
  • MaterialPolystyrene
  • SterileNo
  • DescriptionPS Sampling Spoons, 0.25 mL Capacity; 25/Pk
Reviews
Q&A