Scienceware F18876-0020 Floating Cryotube Rack (holds 20 X 2 mL Cryotubes)

Representative image only.

Scienceware F18876-0020 Floating Cryotube Rack (holds 20 X 2 mL Cryotubes)

Scienceware | Mfr: F18876-0020| Item: 0627402

Racks float in water baths and stand on benchtops

₹9,500.00INR / Each

₹9,500.00INR / Each
Product Description

More About this Item

The filled rack unloads easily by pressing the rack down against the bench until the legs touch, releasing the tubes.

Specifications
  • Tubes Held20
  • Tube Size1.0 to 2.0 mL
  • ColorClear
  • DescriptionFloating Cryotube Rack (holds 20 X 2 mL Cryotubes)
Reviews
Q&A