Scienceware Circular Pipette Stand, Polypropylene, Fixed, Holds 28 Pipettes
Representative image only.
Scienceware Circular Pipette Stand, Polypropylene, Fixed, Holds 28 Pipettes
Scienceware | Mfr: 18955-0000| Item: 0621710
Space saving pipette storage
₹10,100.00INR / Each
₹10,100.00INR / Each
Product Description
Key Features
  • Constructed of durable polypropylene
  • Pipette tips will not contact table surface
More About this Item
Circular pipette support stands provide safe and orderly storage with a space saving design.
Choose from a stationary or rotating stand. The stationary stand holds 28 pipettes, ten 14 mm and eighteen 11 mm diameter pipettes. The rotating stand holds up to 94 pipettes and rotates on a central vertical axis for convenient selection.
Polypropylene plastic stand has a two-section top plate—one with 10 holes, 14mm in diameter, the other with 18 holes, 11mm in diameter. Holes in the lower plate are 4mm in diameter and support the tips of the pipettes as they are held in the stand.
Specifications
  • Tubes Held28
  • Length (in)7
  • Height (in)7
  • MaterialPolypropylene(PP)
  • TypeFixed Stand
  • DescriptionCircular Pipette Stand, Polypropylene, Fixed, Holds 28 Pipettes