Scienceware 28461-1000 graduated polypropylene cylinder with handle, 1000 mL

Representative image only.

Scienceware 28461-1000 graduated polypropylene cylinder with handle, 1000 mL

Scienceware | Mfr: 28461-1000| Item: 0613710

₹9,300.00INR / Each

for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹9,300.00INR / Each

Buy It Together

Current Item

Scienceware 28461-1000 graduated polypropylene cylinder with handle, 1000 mL
Scienceware 28461-1000 graduated polypropylene cylinder with handle, 1000 mL
0613710

₹9,300.00INR / Each
+ =
Total Price 23800.00 ADD 2 ITEMS TO CART
Product Description

More About this Item

The handle on these polypropylene cylinders provides a solid grip, making pouring safe and easy. The hand opening on the handle measures 3-1/2'' x 1-1/8''. Cylinders have a double scale with raised graduations for easy reading. Sturdy hexagonal base. Steam autoclavable at 250degF (121degC). Bel Art.

Specifications
  • MaterialPolypropylene
  • Capacity (mL)1000
  • Graduation Interval (mL)10
  • Descriptiongraduated polypropylene cylinder with handle, 1000 mL
Reviews
Q&A