Scienceware 20320-1660 PTFE Joint Stirrer Bearing for Standard Taper Size 24/40; 1/Pk
Representative image only.
Scienceware 20320-1660 PTFE Joint Stirrer Bearing for Standard Taper Size 24/40; 1/Pk
Scienceware | Mfr: 20320-1660| Item: 0613985
Fits stirring rods with 10mm diameter
₹46,800.00INR / Each
₹46,800.00INR / Each
Product Description
Includes
Extra bushing
Key Features
  • Threaded PP locking compression cap
More About this Item
The joint stirrer bearing is ideal for use with 10-mm OD stirring rods. Use O-ring for fine adjustment of rod tension. Features a threaded PP locking compression cap. Removable bushing fits inside adapter (extra bushing included).
Specifications
  • Taper Size24/40
  • Port Size (ID) (mm)10
  • MaterialPolytetrafluoroethylene (PTFE)
  • DescriptionPTFE Joint Stirrer Bearing for Standard Taper Size 24/40; 1/Pk