Savillex 450-50-3 PTFE 50-mm Membranes; porosity, 1-2µ; thickness, 10 mil; 50/pack
Representative image only.
Savillex 450-50-3 PTFE 50-mm Membranes; porosity, 1-2µ; thickness, 10 mil; 50/pack
Savillex | Mfr: 450-50-3| Item: 0662130
₹168,000.00INR / Pkg of 10
₹168,000.00INR / Pkg of 10
Product Description
More About this Item
Wrench set is for assembly and disassembly, and PTFE membranes are available in different porosities and thicknesses.
Specifications
  • MaterialHydrophobic PTFE
  • Pore Size (µm)1 to 2
  • Diameter (mm)50
  • Thickness (mil)10
  • DescriptionPTFE 50-mm Membranes; porosity, 1-2µ; thickness, 10 mil; 10/pack