SAS Super 100 Air Sampler Tripod and Adapter
Representative image only.
SAS Super 100 Air Sampler Tripod and Adapter
SAS | Mfr: 42704| Item: 3368750
Optimize performance of your SAS Super 100 Digital Microbial Air Samplers
₹248,000.00INR / Each
₹248,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
These accessories are compatible with models 33687-02 and -12 SAS Super 100 Digital Microbial Air Samplers.
The 33687-52 petri head adapter is used with 100 mm x 15 mm petri dishes.
The 33687-50 tripod includs an adapter for quick release of your air sampler.
Includes adapter for quick release of air sampler
1 Year Warranty
Specifications
  • Compatible With33687-02, 33687-12
  • DescriptionSuper 100 Air Sampler Tripod and Adapter