Sartorius YWP09-3 Weighing Pan for Cubis® II High-Capacity Micro Balance, 50 mm dia

Representative image only.

Sartorius YWP09-3 Weighing Pan for Cubis® II High-Capacity Micro Balance, 50 mm dia

Sartorius | Mfr: YWP09-3| Item: 3974361

Optimize performance of your Cubis® II balances

₹102,000.00INR / Each

₹102,000.00INR / Each
Product Description

Key Features

  • Slotted
  • Compatible with high-capacity micro balances balances

More About this Item

Replacement parts and accessories for your Sartorius Cubis II series balances help save time and offer more accurate results.

Please Note: The balance is sold separately.

Specifications
  • Accessory TypeWeighing pan
  • Diameter (mm)50
  • DescriptionWeighing Pan for Cubis® II High-Capacity Micro Balance, 50 mm dia
Reviews
Q&A