Sartorius YSH30-3 Filter Holder for Cubis® II High-Capacity Micro Balance, 150 mm dia

Representative image only.

Sartorius YSH30-3 Filter Holder for Cubis® II High-Capacity Micro Balance, 150 mm dia

Sartorius | Mfr: YSH30-3| Item: 3974356

Optimize performance of your Cubis® II balances

₹291,000.00INR / Each

₹291,000.00INR / Each
Product Description

Key Features

  • Allows easy weighing of filters up to a diameter of 150 mm
  • Compatible with high-capacity micro balances balances

More About this Item

Replacement parts and accessories for your Sartorius Cubis II series balances help save time and offer more accurate results.

Specifications
  • Accessory TypeFilter holder
  • Diameter (mm)150
  • DescriptionFilter Holder for Cubis® II High-Capacity Micro Balance, 150 mm dia
Reviews
Q&A