Sartorius YSH26-3 Weighing Paper Holder for Cubis® II High-Capacity Micro Balance, 60 x 40 mm
Representative image only.
Sartorius YSH26-3 Weighing Paper Holder for Cubis® II High-Capacity Micro Balance, 60 x 40 mm
Sartorius | Mfr: YSH26-3| Item: 3974358
Optimize performance of your Cubis® II balances
₹259,000.00INR / Each
₹259,000.00INR / Each
Product Description
Key Features
  • Allows easy and secure positioning of weigh boats or papers
  • Reduces interferences caused by electrostatic influences
  • Compatible with high-capacity micro balances
More About this Item
Replacement parts and accessories for your Sartorius Cubis II series balances help save time and offer more accurate results.
Specifications
  • Accessory TypeWeighing paper holder
  • DescriptionWeighing Paper Holder for Cubis® II High-Capacity Micro Balance, 60 x 40 mm