Sartorius YSH12-3 Stent Holder for Cubis® II High-Capacity Micro Balance, 38 mm

Representative image only.

Sartorius YSH12-3 Stent Holder for Cubis® II High-Capacity Micro Balance, 38 mm

Sartorius | Mfr: YSH12-3| Item: 3974357

Optimize performance of your Cubis® II balances

₹356,000.00INR / Each

₹356,000.00INR / Each
Product Description

Key Features

  • Designed to position coronary stents up to 38 mm
  • Compatible with high-capacity micro balances

More About this Item

Replacement parts and accessories for your Sartorius Cubis II series balances help save time and offer more accurate results.

Specifications
  • Accessory TypeStent holder
  • Diameter (mm)38
  • DescriptionStent Holder for Cubis® II High-Capacity Micro Balance, 38 mm
Reviews
Q&A