Sartorius YSH02-3 Sample Holder for Cubis® II High-Capacity Micro Balance, up to 50 mL

Representative image only.

Sartorius YSH02-3 Sample Holder for Cubis® II High-Capacity Micro Balance, up to 50 mL

Sartorius | Mfr: YSH02-3| Item: 3974350

Optimize performance of your Cubis® II balances

₹399,000.00INR / Each

₹399,000.00INR / Each
Product Description

Key Features

  • Allows easy positioning of long neck flasks or vials up to 50 mL
  • Compatible with high-capacity micro balances

More About this Item

Replacement parts and accessories for your Sartorius Cubis II series balances help save time and offer more accurate results.

Specifications
  • Accessory TypeSample holder
  • Capacity (mL)50
  • DescriptionSample Holder for Cubis® II High-Capacity Micro Balance, up to 50 mL
Reviews
Q&A