Sartorius YDK04 Specific Gravity Kit for 10mg Secura, Quintix, Practum Balances
Representative image only.
Sartorius YDK04 Specific Gravity Kit for 10mg Secura, Quintix, Practum Balances
Sartorius | Mfr: YDK04| Item: 1122960
Expand the uses of your balance with the below-balance weighing hook
₹284,000.00INR / Each
₹284,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
A hook is available underneath all balances for weighing beneath the balance. This is particularly useful in applications such as determining the measurment of large pieces.
1 Year Warranty
Specifications
  • DescriptionSpecific Gravity Kit for 10mg Secura, Quintix, Practum Balances