Sartorius YDK01 Density Determination Kit
Representative image only.
Sartorius YDK01 Density Determination Kit
Sartorius | Mfr: YDK01| Item: 2575100
₹431,000.00INR / Each
₹431,000.00INR / Each
Buy It Together
Current Item
Sartorius YDK01 Density Determination Kit
Sartorius YDK01 Density Determination Kit
2575100
₹431,000.00INR / Each
+
Cole-Parmer RS-232 to USB Cable
1010083
₹9,900.00INR / Each
+ =
Total Price 1033900.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
two beakers (76 and 55 mm), bar frame, thermometer with retainer clip, glass plummet, sieve for immersing samples, sample holder, metal platform, gasket for ME models, and three adapters.
Key Features
  • Denisty kit for analytical balances
  • 1 year warranty
More About this Item
Use with the buoyancy technique, displacement principle, or the pycnometer method to determine density of solid or liquid samples. Kit includes weighing pans and plummets to easily adapt to your application. Sample holders let you measure in air or in a medium causing buoyancy. Special design prevents air bubbles from adhering so results won't be distorted. Specially shaped sieve lets you completely immerse your sample to weigh a substance with a density less than that of the liquid causing buoyancy. Compatible with balances with different readability ranges — simply exchange the pan adapter. Use with most Sartorius analytical balances.
1 Year Warranty
Specifications
  • DescriptionDensity Determination Kit