Sartorius Vivaspin 20 Centrifugal Concentrators, PES Membrane 30,000 Da; 12/Pk
Representative image only.
Sartorius Vivaspin 20 Centrifugal Concentrators, PES Membrane 30,000 Da; 12/Pk
Sartorius | Mfr: VS2021| Item: 3622474
Broad range of process alternatives for the filtration and concentration of biological samples
₹42,700.00INR / Pkg of 12
₹42,700.00INR / Pkg of 12
Product Description
Key Features
  • Disposable ultrafiltration devices concentrate and purify biological samples to concentrations over 100x in a single spin!
  • Concentrators feature twin vertical membranes—increased surface area provides faster filtration
  • Polyethersulfone membrane is excellent for most solutions when recovery is most important
More About this Item
Vivaspin 20 handles up to 20 ml in swing bucket centrifuges and 14 ml in fixed angle rotors accepting 50 ml centrifuge tubes.
Specifications
  • Max Molecular Weight Cut Off (Mwco) (Dalton)30000
  • Max Volume Range (µL)20
  • Min Volume Range (mL)5
  • DescriptionCentrifugal Concentrators, PES Membrane 30,000 Da; 12/Pk