SalvisLab 31F04015 Exhaust air Adapter for Thermocenter ovens 52403-00 and 52403-05
Representative image only.
SalvisLab 31F04015 Exhaust air Adapter for Thermocenter ovens 52403-00 and 52403-05
SalvisLab | Mfr: 31F04015| Item: 5240070
₹104,000.00INR / Each
₹104,000.00INR / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
From shelving to replacement sensors and communication cables, find everything your need to make the most of your Salvislab oven.
Exhaust air adapter for Salvis Thermocenter ovens 52403-00 and 52403-05. Required for proper air ventilation when stacking two ovens using the stacking adapter part number 52400-80.
Two year warranty
Specifications
  • DescriptionExhaust air Adapter for Thermocenter ovens 52403-00 and 52403-05
  • Warranty2 Years