Saint Gobain Pure-Fit® SIB® Sanitary to Hose Barb Fitting, Straight Adapter, Polypropylene, 3/4" Mini x 5/8"; 25/Pk
Representative image only.
Saint Gobain Pure-Fit® SIB® Sanitary to Hose Barb Fitting, Straight Adapter, Polypropylene, 3/4" Mini x 5/8"; 25/Pk
Saint Gobain | Mfr: PFMS625PPAF| Item: 5014349
Smooth inner bore ensures seamless fluid transition between tubing and fittings
₹42,200.00INR / Pkg of 25
₹42,200.00INR / Pkg of 25
Product Description
Key Features
  • Unique internal design eliminates dead space
  • No areas of entrapment—prohibiting a potential for bacterial growth
  • Use with Pure-Fit and BarbLock® retainers for leak-free connections
  • Easily transition from sanitary fitting to a barb connection
  • Steriliza by gamma irradiation
  • Animal-derived component free and BPA free
  • Manufactured and packaged in Class 8 cleanroom
  • USP <88> Class VI and USP <87> compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • 3/4" Mini x 5/8" hose barb
  • Manufacturer defect warranty
More About this Item
Eliminate dead space and entrapment, a potential breeding ground for bacterial growth, with smooth inner bore (SIB) fittings. SIB fittings provide a seamless, “zero” transition between tubing and fittings, maintaining complete fluid integrity. Straight adapters allow easy transition from a sanitary fitting system to a barbed connection. All fittings feature a 3/4" mini fitting and a barbed fitting—order a gasket and clamp to complete your sanitary connection. Use with Pure-Fit or BarbLock retainer assembly that provides 360° compression to ensure a leak-free connection.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolypropylene
  • Sterilization MethodGamma irradiation
  • Number Of Ports2
  • Connection TypeMini x Hose Barb
  • Barb Size / Tubing Id (Port 2) (mm)15.9
  • Barb Size / Tubing Id (Port 2) (in)5/8
  • Sanitary Clamp Size (Port 1) (InchShort)3/4
  • Flow PatternStraight
  • Max Temperature (° F)200
  • Max Temperature (° C)93
  • Max Operating Pressure (PSI)250
  • Regulatory ApprovalsADCF;USP
  • DescriptionPure-Fit® SIB® Sanitary to Hose Barb Fitting, Straight Adapter, Polypropylene, 3/4" Mini x 5/8"; 25/Pk
  • WarrantyManufacturer defect