Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 6"L 3-Port; 500 mL, 20/Pk
Representative image only.
Saint Gobain 2-D Sterile Single-Use BioProcess Bags, 6"L 3-Port; 500 mL, 20/Pk
Saint Gobain | Mfr: BP0500M| Item: 1594608
Readily integrate into your single-use process applications
Call For Price
Call For Price
Product Description
Key Features
  • Free of any animal-derived components for greater process purity
  • Manufactured in a Class 10,000 facility
  • Needle-free sampling port for added safety
  • Integrated handle on 2- through 20-L sizes
More About this Item
Replace traditional rigid-walled plastic and glass tubes, flasks, bottles, and carboys with these sterile, single-use liquid handling systems. The collapsible containers minimize the risk of cross-contamination and reduce cleaning and turnaround time. Offering outstanding biocompatibility, chemical compatibility, barrier and strength properties, these bags meet your most demanding bioprocessing applications.
Multilayer film structure is optimized for bioprocess applications. Film materials are consistent throughout the product range; with low-density polyethylene (LDPE) as the fluid contact material and ethyl vinyl alcohol (EVOH) as the gas barrier layer. Products are certified to be free of animal-derived components and pass USP Class VI medical-grade testing.
Bags maintain integrity at temperatures ranging from –80 to 60°, making them suitable for packaging, ambient, and cryogenic storage of a variety of liquids including media, buffers, process liquids, and biological products. Sterilized by gamma radiation, ports are capped and covered, and systems are double-bagged for integrity.
Specifications
  • Description2-D Sterile Single-Use BioProcess Bags, 6"L 3-Port; 500 mL, 20/Pk