Safety Spark Laboratory Lighter with 5 Flints
Representative image only.
Safety Spark Laboratory Lighter with 5 Flints
| Mfr: 3021| Item: 3613650
Sure, safe ignition for your bunsen burner
₹1,600.00INR / Each
₹1,600.00INR / Each
Buy It Together
Current Item
Safety Spark Laboratory Lighter with 5 Flints
Safety Spark Laboratory Lighter with 5 Flints
3613650
₹1,600.00INR / Each
+
Flint for Safety Spark Laboratory Lighter; 5/Pk
3613655
₹610.00INR / Pkg of 5
=
Total Price 2210.00 ADD 2 ITEMS TO CART
Product Description
More About this Item
Use this well-constructed, safety spark lighter for all your burner ignition needs. Special spark hood protects the flint. Gas pocket forms beneath the hood, ensuring instant lighting.
Lighter incorporates new larger, renewable flint that produces up to 1500 ignitions..
Specifications
  • DescriptionSafety Spark Bunsen Burner/Laboratory Lighter with 5 Flints