Safeskin 03040-CS PURPLE NITRILE Exam Gloves, (S) 24.13 cm (9.5"), 1000/ Case, Purple

Representative image only.
Safeskin Mfr # 03040-CS – Item # WW-33466-04
Purple color easily distinguishes these gloves from latex
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.

PRODUCT OPTIONS

₹11,400.00INR / Case of 1000

SPECS & DESCRIPTION

Specifications & Description

  • MaterialNitrile
  • SizeSmall
  • Length (in)9 1/2
  • Length (cm)24
  • ColorPurple
  • SterileNo
  • DescriptionPURPLE NITRILE Exam Gloves, (S) 24.13 cm (9.5\"), 1000/ Case, Purple

Key Features

  • Due to the high demand for personal safety products, cancellations and returns will be restricted.

More About this Item

These 100% nitrile gloves are soft, latex-free, powder-free, and nonsticky. Low chemical content reduces risk of allergic reactions. Textured fingertips enhance wet or dry grip. Beaded cuff. Ambidextrous.

Compliance: ASTM D 6319, ISO 9002.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat