Rubbermaid 1933 NSF-Approved High-Heat Spatulas, PP/Rubber, 9-1/2"L
Representative image only.
Rubbermaid 1933 NSF-Approved High-Heat Spatulas, PP/Rubber, 9-1/2"L
Rubbermaid | Mfr: 10613377| Item: 0627815
₹11,900.00INR / Pkg of 6
₹11,900.00INR / Pkg of 6
Product Description
Key Features
  • 1 year warranty
More About this Item
Scooping, spooning, and spreading have never been easier with these spoon-shaped spatulas. The sturdy polypropylene (PP) handle is permanently bonded to a flexible rubber blade. Spatulas withstand temperatures to 212°F (100°C). NSF-approved.
1 Year Warranty
Specifications
  • Overall Length (in)9 1/2
  • Overall Length (cm)24.13
  • MaterialPolypropylene, Rubber
  • SterileNo
  • DescriptionNSF-Approved High-Heat Spatulas, PP/Rubber, 9-1/2"L