Rite in the Rain 550F Bound Environmental Notebook
Representative image only.
Rite in the Rain 550F Bound Environmental Notebook
Rite in the Rain | Mfr: 550F| Item: 0657000
Sturdy polydura covers for durability
₹10,000.00INR / Each
₹10,000.00INR / Each
Product Description
Key Features
  • All-weather paper can be written even when wet
  • Secure sewn-in pages which lay flat when open
  • Rounded corners
  • Reference material and numbered pages meet US EPA guidelines
  • Square grids on front page, rectangular grids on back page
  • 156 numbered pages (78 sheets)
More About this Item
The Rite in the Rain bound books have sturdy polydura covers. Books measure 4-3/4" x 7-1/2" and have 156 numbered pages (78 sheets). All-weather paper can be written on even when wet; use pencil or crayon when wet, ballpoint pen or felt-tip pen when dry. Sewn-in pages stay securely in place and lay flat when open.
The environmental book meets US EPA guidelines and contains ten helpful reference pages with a field page pattern, imperial & metric rulers, conversion table, and map scale.
Specifications
  • Number of Pages156
  • ColorYellow
  • DescriptionBound Environmental Notebook; 1/Pk