Rex Gauge OS-3 Accessory Test stand for micro A durometers
Representative image only.
Rex Gauge OS-3 Accessory Test stand for micro A durometers
Rex Gauge | Mfr: OS-3| Item: 5979264
Optimize performance of your Rex Gauge durometer
₹375,000.00INR / Each
₹375,000.00INR / Each
Product Description
Key Features
  • Test stands improve ease of use
  • 1 year warranty
More About this Item
Test stands provide a convenient and accurate way to perform repeated hardness tests. Use stands to test pressure uniformly for compliance with ASTM D-2240 and eliminate human error.
The AC adapter is suitable for use with 220 VAC power input.
1 Year Warranty
Specifications
  • DescriptionAccessory Test stand for micro A durometers