Replica-Plating Tool for 90 to 100-mm Petri Dishes
Representative image only.
Replica-Plating Tool for 90 to 100-mm Petri Dishes
| Item: 1421000
Accurately replicate colonies grown on petri dishes
₹74,100.00INR / Each
₹74,100.00INR / Each
Buy It Together
Current Item
Replica-plating tool for 90 to 100-mm petri dishes
Replica-plating tool for 90 to 100-mm petri dishes
1421000
₹74,100.00INR / Each
+
Replacement velveteen squares for 14210-00
1421050
₹72,500.00INR / Box of 36
=
Total Price 146600.00 ADD 2 ITEMS TO CART
Product Description
Includes
PVC cylinder, ocking aluminum ring, and twelve sterile velveteen squares.
Key Features
  • Compatible with 90- to 100-mm petri dishes
More About this Item
Use this economical device to accurately replicate colonies grown on 90- to 100-mm petri dishes. A locking aluminum ring (102 mm dia) secures a sterile velveteen square onto a PVC cylinder for easy control of the contact surface.
To replicate colonies, invert master petri dish over device to transfer colony pattern to fabric, remove, and lower device onto fresh agar plate. Disinfect between uses with a brief rinse in ethanol or chlorine bleach. Cylinder is 55 mm H with 80-mm and 88-mm dia ends.
Specifications
  • MaterialPVC, aluminum, and velveteen
  • Height (mm)55
  • Diameter (mm)102
  • DescriptionReplica-plating tool for 90 to 100-mm petri dishes