Replacement Heating Element for Bacti-Cinerator III; 120 VAC

Representative image only.

Replacement Heating Element for Bacti-Cinerator III; 120 VAC

Benchmark Scientific| Item: 0185018

Keep your BactiZapper running with replacement heating elements.

₹106,000.00INR / Each

₹106,000.00INR / Each
Product Description

Key Features

  • This is a replacement for the Bacti-Cinerator only
  • 6 month warranty
Canadian Standards Association6 Month Warranty
Specifications
  • Power (VAC)120
  • DescriptionReplacement Heating Element for Bacti-Cinerator III; 120 VAC
  • Warranty6 month
Reviews
Q&A