Reichert 13754000 Industrial Fluid Refractometer, 0 to 30 scale (1.3330 to 1.3860 RI)
Representative image only.
Reichert 13754000 Industrial Fluid Refractometer, 0 to 30 scale (1.3330 to 1.3860 RI)
Reichert | Mfr: 13754000| Item: 8100024
Monitor quality of industrial fluids and coolants with ease
₹81,100.00INR / Each
for an additional
₹28,000.00INR / Each
for an additional
₹29,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹81,100.00INR / Each
Buy It Together
Current Item
Reichert 13754000 Industrial Fluid Refractometer, 0 to 30 scale (1.3330 to 1.3860 RI)
Reichert 13754000 Industrial Fluid Refractometer, 0 to 30 scale (1.3330 to 1.3860 RI)
8100024
₹81,100.00INR / Each
+
InnoCal NIST-Traceable Calibration; Refractometer
1710500
₹28,000.00INR / Each
=
Total Price 109100.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Automatic temperature compensation provides accurate readings at any temperature
  • Large, easy-to-read scales and depth of field
  • Portable – no batteries or power cord required
  • Rugged, chemical-resistant housing
  • 1 year warranty
More About this Item
Handheld industrial fluid refractometers designed to quickly determine ratio or concentration of water-soluble industrial fluids and lubricants. Ideal for synthetic lubricants, cutting fluids, and water-soluble lubricants.
Note: Scale does not display refractive index values.
1 Year Warranty
Specifications
  • Resolution0.2 (RI)
  • Accuracy±0.001 (RI)
  • Max Temperature Compensation (° F)100
  • Min Temperature Compensation (° F)60
  • Max Temperature Compensation (° C)38
  • Min Temperature Compensation (° C)15
  • DescriptionIndustrial Fluid Refractometer, 0 to 30 scale (1.3330 to 1.3860 RI)