Raven Environmental B-10101-19 Replacement Centrifuge Tubes for 05477-10 and -20
Representative image only.
Raven Environmental B-10101-19 Replacement Centrifuge Tubes for 05477-10 and -20
Raven Environmental | Mfr: B-10101-19| Item: 0547750
Provides easy-to-read measurements
₹35,800.00INR / Pkg of 12
₹35,800.00INR / Pkg of 12
Product Description
Key Features
  • Impact resistant
  • 3 month warranty
More About this Item
These 19 mm polycarbonate tubes are impact resistant with a conical, tapered design. Shows percent volume of compacted solds for easy measurement.
3 Month Warranty
Specifications
  • Compatible With05477-10, -20
  • DescriptionReplacement centrifuge tubes for 05477-10 and -20