Radiation Alert WIPE TEST PLATE Wipe Test Plate for 8191010
Representative image only.
Radiation Alert WIPE TEST PLATE Wipe Test Plate for 8191010
Radiation Alert | Mfr: WIPE TEST PLATE| Item: 8191060
₹11,700.00INR / Each
₹11,700.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Stainless steel plate positions wipe directly under sensor at a fixed distance of 1 cm for gross wipe counting; slides out easily for general surveying or decontamination.
1 Year Warranty
Specifications
  • DescriptionWipe Test Plate for 8191010