Radiation Alert THE-CHARGER Radiation Dosimeter Charger
Representative image only.
Radiation Alert THE-CHARGER Radiation Dosimeter Charger
Radiation Alert | Mfr: THE-CHARGER| Item: 8192200
Charge your dosimeter without batteries
₹111,000.00INR / Each
₹111,000.00INR / Each
Buy It Together
Current Item
Radiation Alert THE-CHARGER Radiation Dosimeter Charger
Radiation Alert THE-CHARGER Radiation Dosimeter Charger
8192200
₹111,000.00INR / Each
+
Radiation Alert Personal Radiation Dosimeter, 0 to 200 Mr
8192210
₹52,300.00INR / Each
=
Total Price 163300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 15,000 volt piezoelectric generator
  • 1 year warranty
More About this Item
The handheld Charger can charge a variety of direct-reading dosimeters by simply squeezing the lever of a generator. The Charger requires no batteries. By pulling a trigger, a dosimeter is easily placed in or removed from the charger. The clamping action holds the dosimeter for you.
1 Year Warranty
Specifications
  • DescriptionRadiation Dosimeter Charger