QRP 70G Heat Gloves, 12 inches

Representative image only.

QRP 70G Heat Gloves, 12 inches

QRP | Mfr: 70G| Item: 3368931

₹146,000.00INR / Pair of 1

₹146,000.00INR / Pair of 1
Product Description

Key Features

  • 3 month warranty

More About this Item

Ideal for use with diffusion furnaces, burn-in ovens, ceramics, and food processing and handling. These flexible ESD gloves made with PolyTuff® silicon formulation protect you in wet or dry and withstand temperatures from -78.5 to 450°F (-61.4 to 232°C). Resistant to solders and fluxes, gloves are free of PCB, asbestos, and fiberglass hazards. Packaged for immediate cleanroom use.
Compliance: Class 100

3 Month WarrantyConformite Europeenne
Specifications
  • MaterialPolytuff®
  • SizeLarge
  • Length (in)12
  • Length (cm)30.5
  • ColorWhite
  • ComplianceClass 100
  • SterileNo
  • DescriptionHeat Gloves, 12 inches
  • Warranty3 month
Reviews
Q&A