QLA Transdermal Patch Holder with Watch glass, 17 mesh PTFE screen and PVDF Clips
Representative image only.
QLA Transdermal Patch Holder with Watch glass, 17 mesh PTFE screen and PVDF Clips
| Mfr: APPFIVE-HR| Item: 4130100
Transdermal patch holder secures sample for testing
₹39,700.00INR / Each
₹39,700.00INR / Each
Product Description
Key Features
  • Tests patches up to 90 mm in diameter Compatible with all brands of dissolution baths Made of nonreactive materials
More About this Item
The QLA transdermal patch holder is a borosilicate watch glass with a 17 mesh polytetrafluoroethylene (PTFE) screen, held together by polyvinylidene fluoride (PVDF) clips.
.
Not available for sale in the United States.
Specifications
  • MaterialPTFE screen with PVDF clips
  • Mesh Size17
  • Diameter (cm)9