QLA Time Release Capsule Sinker; 316 Stainless Steel
Representative image only.
QLA Time Release Capsule Sinker; 316 Stainless Steel
| Mfr: CAPWHT-TR| Item: 4130294
Capsule sinkers improve reproducibility
₹13,800.00INR / Each
₹13,800.00INR / Each
Product Description
Key Features
  • Keep floating tablets from sticking to sides of vessels
  • 0.211" (5.4 mm) ID
More About this Item
Sinkers help to ensure tablets dissolve at more reproducible rates and keep them from sticking to the sides of vessels. Select from spring style, basket style, three-prong style, and others.
Specifications
  • Material316 stainless steel
  • Diameter (InchShort)13/64
  • Diameter (mm)5.4
  • DescriptionTime Release Capsule Sinker; 316 Stainless Steel