QLA Standard Vessel for Pharmatest, 1000 mL, Amber Glass; Serialized

Representative image only.

QLA Standard Vessel for Pharmatest, 1000 mL, Amber Glass; Serialized

| Mfr: GLA900-APT| Item: 4130626

QLA vessels provide more precise and consistent dissolution results

₹87,600.00INR / Each

₹87,600.00INR / Each
Product Description

Key Features

  • 1000 mL capacity
  • Standard vessel
  • Amber glass is ideal for light-sensitive compounds
  • A unique serial number is acid etched on the vessel

More About this Item

Vessels are manufactured using a unique molding process to provide a precise and consistent inside diameter and spherical radius to meet the physical dimensions of the USP.

Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Footed plastic vessels are an economical alternative to glass. They feature feet which allow them to stand upright independently.

.

Specifications
  • Capacity (mL)1000
  • MaterialGlass
  • Bottle ColorAmber
  • Special FeaturesStandard
  • DescriptionStandard Vessel for Pharmatest, 1000 mL, Amber Glass; Serialized
Reviews
Q&A