QLA Standard Vessel for Erweka, 2000 mL, Clear Glass; Serialized

Representative image only.

QLA Standard Vessel for Erweka, 2000 mL, Clear Glass; Serialized

| Mfr: GLA02K-EW| Item: 4130603

QLA vessels provide more precise and consistent dissolution results

₹88,500.00INR / Each

₹88,500.00INR / Each
Product Description

Key Features

  • Achieve reproducible results with precision molded vessels
  • Clear glass
  • 2000 mL capacity
  • Serialized

More About this Item

Standard glass vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Footed plastic vessels are an economical alternative to glass. They feature feet which allow them to stand upright independently.

Glass vessels offer good thermal conductivity as well as excellent resistance to scratching. A unique serial number is acid etched on the vessel.

.

Specifications
  • Capacity (mL)2000
  • MaterialGlass
  • Bottle ColorClear
  • Special FeaturesStandard
  • DescriptionStandard Vessel for Erweka, 2000 mL, Clear Glass; Serialized
Reviews
Q&A