QLA Spring Clip Style Basket Shaft for Hanson SR8-Plus, 16"L, 316 SS; Serialized
Representative image only.
QLA Spring Clip Style Basket Shaft for Hanson SR8-Plus, 16"L, 316 SS; Serialized
| Mfr: BSKSHT-16| Item: 4130181
QLA Shafts for dissolution testing with your Hanson apparatus
₹60,300.00INR / Each
₹60,300.00INR / Each
Buy It Together
Current Item
QLA Spring Clip Style Basket Shaft for Hanson SR8-Plus, 16"L, 316 SS; Serialized
QLA Spring Clip Style Basket Shaft for Hanson SR8-Plus, 16"L, 316 SS; Serialized
4130181
₹60,300.00INR / Each
+
QLA Basket Holder, Universal Compatibility; 12 Position
4130169
₹49,600.00INR / Each
=
Total Price 109900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Made of 316 electropolished stainless steel All shafts are laser marked with unique serial numbers
More About this Item
These QLA shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Hanson test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
.
Specifications
  • Material316 stainless steel
  • Length (in)16
  • DescriptionSpring Clip Style Basket Shaft for Hanson SR8-Plus, 16"L, 316 SS; Serialized