QLA Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 21"L, 316 SS; Serialized
Representative image only.
QLA Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 21"L, 316 SS; Serialized
| Mfr: BSKSHT-21| Item: 4130188
QLA Shafts for dissolution testing with your Agilent/VanKel/Varian apparatus
₹68,500.00INR / Each
₹68,500.00INR / Each
Product Description
Key Features
  • Made of 316 electropolished stainless steel All shafts are laser marked with unique serial numbers
  • Spring clip style
  • 21" long
More About this Item
These QLA shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Agilent/VanKel/Varian test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness and durability. Shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
.
Specifications
  • Material316 stainless steel
  • Length (in)21
  • Length (mm)533
  • DescriptionSpring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 21"L, 316 SS; Serialized