QLA Spiral Capsule Sinker, Orange PTFE-Coated Music Wire; 6.5 coils
Representative image only.
QLA Spiral Capsule Sinker, Orange PTFE-Coated Music Wire; 6.5 coils
| Mfr: CAPWHT-LGMT| Item: 4130290
Capsule sinkers improve reproducibility
₹27,900.00INR / Each
₹27,900.00INR / Each
Product Description
Key Features
  • Keep floating tablets from sticking to sides of vessels
  • 0.41" (10.4 mm) ID
  • Orange PTFE-coated music wire
  • 6.5 coils
More About this Item
Sinkers help to ensure tablets dissolve at more reproducible rates and keep them from sticking to the sides of vessels. Select from spring style, basket style, three-prong style, and others.
Specifications
  • MaterialPTFE-coated music wire
  • Diameter (InchShort)13/32
  • Diameter (mm)10.4
  • DescriptionSpiral Capsule Sinker, Orange PTFE-Coated Music Wire; 6.5 coils